missing translation for 'onlineSavingsMsg'
Learn More

Garcin™ Garcin™ Bactinyl Wipers

Product Code. 11862623
Click to view available options
Description:
Odorless
Scented
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 11862623

Brand: Garcin™ LB920630

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Clean and disinfect surfaces and instruments with Garcin™ Bactinyl Wipers, which are general purpose wipers for general sanitation purposes.

  • Fragranced or odorless
  • Rechargeable dispenser box with reclosable case
  • Active substances include peroxides, quaternary ammoniums, ethanol
  • Mineral-certified impregnation solution

Specifications

Product Type Wipers
Length (Metric) 13 cm
Width (Metric) 20 cm
Description Odorless
Packaging Quantity 12 Pk.
compliance-icons
Product Identifier
  • bactinyl lingettes disinfectant indoor
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, didecyldimethylammonium chloride, benzalkonium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.