Learn More
Garcin™ Garcin™ Bactinyl Wipers
Product Code. p-8000714
Click to view available options
Description:
Odorless
Scented
Unit Size:
Pack of 12
Product Code. 11862623
Brand: Garcin™ LB920630
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Clean and disinfect surfaces and instruments with Garcin™ Bactinyl Wipers, which are general purpose wipers for general sanitation purposes.
  • Fragranced or odorless
  • Rechargeable dispenser box with reclosable case
  • Active substances include peroxides, quaternary ammoniums, ethanol
  • Mineral-certified impregnation solution
Specifications
Product Type Wipers
Length (Metric) 13 cm
Width (Metric) 20 cm
Description Odorless
Packaging Quantity 12 Pk.
compliance-icons
Product Identifier
  • bactinyl lingettes disinfectant indoor
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, didecyldimethylammonium chloride, benzalkonium chloride