Learn More
Invitrogen™ Gardnerella vaginalis Monoclonal Antibody (4971)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547540
Description
Specificity: Specific for Gardnerella vaginalis. No cross reactivity with: Candida albicans, Chlamydia, Group B Strep, E. coli, Proteus sp., N. gonorrhoeae, Corynebacteria sp., HSV2, Mycoplasma hominis, Urealplasma urealyticum. Best Pairs (capture/conjugate): Gardnerella vaginalis Monoclonal antibody (Product # MA5-47540)/Gardnerella vaginalis Monoclonal antibody (Product # MA5-47541); Gardnerella vaginalis Monoclonal antibody (Product # MA5-47542)/Gardnerella vaginalis Monoclonal antibody (Product # MA5-47540); Gardnerella vaginalis Monoclonal antibody (Product # MA5-47541)/Gardnerella vaginalis Monoclonal antibody (Product # MA5-47540); Gardnerella vaginalis Monoclonal antibody (Product # MA5-47543)/Gardnerella vaginalis Monoclonal antibody (Product # MA5-47540). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Specifications
| Gardnerella vaginalis | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
| ELISA, Lateral Flow, Immunocytochemistry | |
| 4971 | |
| Unconjugated | |
| G. vaginalis | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.