missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GARP Mouse anti-Human, BV750, Clone: 7B11 (also known as CMSSC-7B11), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747370
This item is not returnable.
View return policy
Description
GARP Monoclonal antibody specifically detects GARP in Human samples. It is validated for Flow Cytometry
Specifications
| GARP | |
| Monoclonal | |
| Brilliant Violet 750 | |
| LRRC32; Garpin; Glycoprotein A repetitions predominant | |
| Recombinant Human LRRC32 | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| 7B11 (also known as CMSSC-7B11) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction