missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GARS Antibody (4E10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42679F
This item is not returnable.
View return policy
Description
GARS Monoclonal specifically detects GARS in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| GARS | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| GARS | |
| Recombinant human GARS (43-289aa) purified from E. coli | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 2617 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 40000000000 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| AP-4-A synthetase, Charcot-Marie-Tooth neuropathy 2D, Charcot-Marie-Tooth neuropathy, neuronal type, D, Diadenosine tetraphosphate synthetase, DSMAV, glycine tRNA ligase, Glycine--tRNA ligase, glycyl-tRNA synthetase, GlyRSEC 6.1.1.14, HMN5, SMAD1CMT2D | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction