missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GARS Monoclonal Antibody (OTI6C5)
GREENER_CHOICE

Product Code. 15976304
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15976304 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15976304 Supplier Invitrogen™ Supplier No. MA526714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GARS Monoclonal Antibody for Western Blot, IHC (P), IP, RIP

GARS is a glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. This protein is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. The protein has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GARS
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation
Classification Monoclonal
Clone OTI6C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GARS1
Gene Accession No. P41250
Gene Alias 6.1.1.14; AIR synthase; AIRS; Ap4A synthetase; AP-4-A synthetase; Charcot-Marie-Tooth neuropathy 2D; Charcot-Marie-Tooth neuropathy, neuronal type, D; CMT2D; Diadenosine tetraphosphate synthetase; DSMAV; EC 2.1.2.29; GAR transformylase; GARS; gars1; GART; GARTF; Gena201; GENA202; glycine tRNA ligase; glycine--tRNA ligase; glycyl-tran synthetase; glycyl-tRNA synthetase; glycyl-tRNA synthetase 1; glycyl-tRNA synthetase-like protein; glyRS; HMN5; metronome; MGC47764; mrn; Nmf249; PAIS; PGFT; PRGS; RGD1559871; Sgrp23; si:dkey-276i5.1; SMAD1; storage granule protein 23; Unknown (protein for IMAGE:7254992); wu:fb02f03
Gene Symbols GARS, GARS1
Host Species Mouse
Immunogen Full length human recombinant protein of GARS produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2617
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.