Learn More
Trajan Scientific and Medical Gas Filter Connecting Unit
The gas filter connecting unit has inlet and outlet connectors for the gas line and the system can be wall-mounted or fixed to a bench. Connecting units come with a 1, 2 or 4 filter capacity and are available for 1/4 in. or 1/8 in. gas lines
Brand: Trajan Scientific and Medical 1035014
Additional Details : Weight : 3.00000kg
Description
- Easily installed into the gas line
- Can be either wall-mounted or fixed to a bench
- Available with a 1, 2 or 4 filter capacity
Specifications
1/4 in. fittings | |
Gas filters | |
1/4 in. |
1-position Connecting Unit | |