Learn More
Trajan Scientific and Medical Gas Filter Connecting Unit
The gas filter connecting unit has inlet and outlet connectors for the gas line and the system can be wall-mounted or fixed to a bench. Connecting units come with a 1, 2 or 4 filter capacity and are available for 1/4 in. or 1/8 in. gas lines
Brand:  Trajan Scientific and Medical 1035018
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 15750959
  • € 283.00 / Each
Estimated Shipment: 12-04-2024
to see stock.
Description
Description
  • Easily installed into the gas line
  • Can be either wall-mounted or fixed to a bench
  • Available with a 1, 2 or 4 filter capacity
Specifications
Specifications
1/8 in. fittings
Gas filters
1/8 in.
1-position Connecting Unit
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only