Learn More
Lenz Laborglasintrumente Gas Inlet Tubes Bent
Product Code. 10224101
Change view
Click to view available options
Standard Taper Joint:
NS 24/29
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
10224101 NS 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10224101 Supplier Lenz Laborglasintrumente Supplier No. 05083003
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Bent air leak tubes (angle: 90°)
  • Built-in length: 250 mm
  • With NS cone and thin capillary (easily to draw by burner)
  • Made of DURAN™ tubing
Specifications
Length (Metric) 250 mm
Standard Taper Joint NS 24/29