Learn More
Trajan™ Gas-Tight Autosampler Syringes
Click to view available options
Volume (Metric):
5 μL
100 μL
Needle Gauge:
22 G
23 G
Connection Type:
Fixed
Removable
For Use With (Equipment):
CTC, Carlo Erba, Fisons Autosampler
Carlo Erba/Fisons/Leap/CTC ASD200 and AS800
PerkinElmer Autosystem
Unit Size:
Each
Description
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
| Needle Gauge | 22 G |
| Diameter (English) Outer Needle | 0.02 in. |
| Diameter (Metric) Outer Needle | 0.72 mm |
| For Use With (Application) | High performance liquid chromatology |
| Length (English) Needle | 2 in. |
| Product Type | Autosampler Syringe |
| Model | 100R-C/T-GT-LC |
| Quantity | Ea. |
| Volume (Metric) | 100 μL |
| Length (Metric) Needle | 51 mm |
| Show More |