Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Cone Tip
Click to view available options
Volume (Metric):
100 μL
250 μL
500 μL
500 mL
Diameter (Metric) Inner Needle:
0.11 mm
0.20 mm
0.32 mm
Diameter (Metric) Outer Needle:
0.47 mm
0.63 mm
Length (Metric) Needle:
57 mm
85 mm
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
Specifications
| Material | PTFE-tipped (Plunger) |
| Needle Gauge | 26 G |
| Diameter (Metric) Inner Needle | 0.20 mm |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Product Type | Gastight |
| Volume (Metric) | 500 μL |
| Length (Metric) Needle | 85 mm |
| Needle Point Style | Cone |
| Connection Type | Fixed |