Learn More
3M™ Gas & Vapor Filter GF22 A2B2
3M™ Gas & Vapor Filter DT-4005E is a DIN 40 threaded, replaceable filter offering A2B2 protection against certain organic vapors and inorganic gases.
Brand: 3M™ 7100265237
132.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.34000kg
Description
- A2B2 protection against certain organic vapors and inorganic gases
- DIN40 threaded connection
- Compatible with 3M™ Reusable Half Mask HF-300 Series, 3M™ Reusable Full Face Masks FF-300 Series and FF-600 Series
- Compatible with 3M™ Powered Air Respirator PF-600E Series
Specifications
Filter | |
3M™ Powered Air Respirator PF-600E Series | |
DT-4005E | |
10/Cs. |