Learn More
Robu Gas-Washing-Bottle, FD 100ml, Por. 4, NS 29/32
Brand:  Robu 40104
Gas-Washing-Bottle, FD 100ml, Por. 4, NS 29/32
Product Code. 10174071
  • ONLINE EXCLUSIVE PRICE
    € 90.00
    € 79.40 / Each
    Save € 10.60
    11% Off
Only null left
Estimated Shipment: 03-02-2026