Learn More
Lenz Laborglasintrumente™ Gas Washing Bottle Head with Filter Disc
Gas washing head for bottles acc. to DIN 12463
Brand: Lenz Laborglasintrumente™ 05510402
Additional Details : Weight : 0.10000kg
Description
- Acc. to DIN 12596
- With cone NS 29/32
- Material: DURAN™ tubing
Specifications
Duran Tubing | |
2 | |
1 |
Olive Connections | |
Gas washing bottles acc. to DIN 12463 |