missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gas1 Antibody (11C47), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22032C
This item is not returnable.
View return policy
Description
Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Gas1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GAS1 | |
| Human recombinant Gas1 | |
| 0.1 mL | |
| 2619 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, ELISA | |
| 11C47 | |
| Western Blot, ELISA | |
| GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1 | |
| Rat | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction