missing translation for 'onlineSavingsMsg'
Learn More

GAS2 Antibody (SR1577), Novus Biologicals™

Product Code. 18667694 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18667694 50 μg 50µL
18640154 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18667694 Supplier Novus Biologicals Supplier No. NBP32261550ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GAS2 Monoclonal antibody specifically detects GAS2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Serum
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR1577
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:100
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias GAS-2, growth arrest-specific 2, growth arrest-specific protein 2, MGC32610
Host Species Rabbit
Immunogen A synthesized peptide derived from human GAS2 (Uniprot #: O43903)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 2620
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.