missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAS2L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582704
This item is not returnable.
View return policy
Description
Immunogen sequence: RLSRVSSPSP ELGTTPASIF RTPLQLDPQQ EQQLFRRLEE EFLANARALE AVASVTPTGP APDPARAPDP PAPDSAYCSS SSSSSSLSVL GGKCG Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 32%.
The Gas2 protein is thought to play a role in apoptosis by acting as a cell death substrate for caspases. Gas2, a component of the microfilament system, is cleaved by a caspase (caspase-3 or caspase-7) at Asparagine 278 during apoptosis. The cleaved form of Gas2 dramatically induces the re-arrangement of the Actin cytoskeleton and causes potent changes in the shape of the affected cells. Gas2 is a cytoskeleton and peripheral membrane protein that co-localizes with Actin fibers at the cell border and along the stress fibers in growth-arrested fibroblasts. Gas2 is mainly membrane-associated, but when hyperphosphorylated it will accumulate at membrane ruffles, indicating Gas2 involvement in that process. Gar22 (Gas2-related protein on chromosome 22), also designated growth arrest-specific protein 2-like 1 (Gas2-like1), belongs to the Gas2 family and contains one CH (calponinhomo-logy) domain. It is thought to be involved in the cross-linking of microtubules and microfilaments.
Specifications
| GAS2L1 | |
| Polyclonal | |
| Unconjugated | |
| GAS2L1 | |
| 4930500E24Rik; D0Jmb1; Gar22; GAS2L1; GAS2-like protein 1; GAS2-related protein; GAS2-related protein on chromosome 22; growth arrest specific 2 like 1; growth arrest-specific 2 like 1; growth arrest-specific protein 2-like 1; TU-71.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10634 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99501 | |
| GAS2L1 | |
| Recombinant protein corresponding to Human GAS2L1. Recombinant protein control fragment (Product #RP-108158). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido