missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAS41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587270
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
GAS41 has high sequence homology to human MLLT1, and yeast and human MLLT3 proteins. Both MLLT1 and MLLT3 proteins belong to a class of transcription factors, indicating that the encoded protein might also represent a transcription factor. This protein is thought to be required for RNA transcription. This gene has been shown to be amplified in tumors.
Specifications
| GAS41 | |
| Polyclonal | |
| Unconjugated | |
| YEATS4 | |
| 4930573H17Rik; B230215M10Rik; GAS41; Glioma-amplified sequence 41; glioma-amplified sequence-41; nuBI1; NuBI-1; NuMA binding protein 1; nuMA-binding protein 1; RGD1305741; YAF9; YEATS domain containing 4; YEATS domain-containing protein 4; Yeats4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 299810, 64050, 8089 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95619, Q9CR11 | |
| YEATS4 | |
| Recombinant full length Human GAS41. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction