missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GAS7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13454767
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13454767 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13454767 Supplier Invitrogen™ Supplier No. PA530274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (92%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS7
Applications Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene GAS7
Gene Accession No. O60861
Gene Alias AW124766; B230343A10Rik; GAS7; GAS-7; Gas7-cb; growth arrest specific 7; growth arrest-specific 7; growth arrest-specific protein 7; KIAA0394; MLL/GAS7; MLL/GAS7 fusion protein
Gene Symbols GAS7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GAS7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.