Learn More
RETSCH Gasket For Grinding Jar
Brand: RETSCH 22.085.0002
Additional Details : Weight : 0.02000kg
Specifications
For 50mL grinding jar | |
10 Pack | |
50 mL Grinding Jar, Stainless Steel |
Gasket | |
Stainless Steel |