missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastric Lipase Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-60930AF750
This item is not returnable.
View return policy
Description
Gastric Lipase Polyclonal specifically detects Gastric Lipase in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Gastric Lipase | |
| Polyclonal | |
| Western Blot | |
| EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271 | |
| Rabbit | |
| 0.1 mL | |
| 8513 | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LIPF | |
| A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-ProtNo. P07098] | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction