missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastric Lipase Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-60930JF585
This item is not returnable.
View return policy
Description
Gastric Lipase Polyclonal antibody specifically detects Gastric Lipase in Human,Mouse samples. It is validated for Western Blot
Specifications
| Gastric Lipase | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Janelia Fluor 585 | |
| EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271 | |
| A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot# P07098] | |
| 0.1 mL | |
| Autophagy, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Endothelial Cell Markers, Glia Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
| 8513 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction