missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Antibody (GAST/2633), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08685AF488
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Gastrin Monoclonal antibody specifically detects Gastrin in Human samples. It is validated for ELISA
Tekniske data
| Gastrin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| GAS, gastrin | |
| Recombinant full-length human Gastrin protein (Uniprot: P01350) | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Cell Biology, Immunology, Signal Transduction | |
| 2520 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| ELISA | |
| GAST/2633 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion