missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrokine 1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97123AF532
This item is not returnable.
View return policy
Description
Gastrokine 1 Polyclonal specifically detects Gastrokine 1 in Human samples. It is validated for ELISA.
Specifications
| Gastrokine 1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Alexa Fluor 532 | |
| 18 kDa antrum mucosa protein, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, Protein CA11 | |
| Produced in rabbits immunized with purified, recombinant Gastrokine 1 (Q9NS71; Asn35-Asn199). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 56287 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction