Learn More
GAT1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20298-1-AP-150UL
This item is not returnable.
View return policy
Description
The GAT1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GAT1. This antibody recognizes human, mouse , rat antigen. The GAT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| GAT1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 6529 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |