missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77115AF594
This item is not returnable.
View return policy
Description
GATA-3 Polyclonal antibody specifically detects GATA-3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| GATA-3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human GATA3. The immunogen is located within amino acids 340 - 390 of GATA3. Amino Acid Squence: KEGIQTRNRKMSSKSKK | |
| 0.1 mL | |
| Cancer, Innate Immunity | |
| 2625 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction