Learn More
GATA-3 Antibody (CL8660) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88917C
Product Code. 30308215
  • € 737.00 / 0.10mL
Description
GATA-3 Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GATA-3
Monoclonal
DyLight 650
131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
This antibody was developed using a synthetic peptide derived from P23771, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Innate Immunity
2625
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL8660
50mM Sodium Borate
Mouse
Protein A purified