missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-3 Antibody (OTI8H4), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70796G
This item is not returnable.
View return policy
Description
GATA-3 Monoclonal specifically detects GATA-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| GATA-3 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GATA3 | |
| Recombinant protein fragment corresponding to amino acids 155-443 of human GATA-3 (NP_002042) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI8H4 | |
| Immunohistochemistry | |
| 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Innate Immunity | |
| 2625.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction