missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-3 Antibody (rGATA3/3870), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20864B
This item is not returnable.
View return policy
Description
GATA-3 Monoclonal antibody specifically detects GATA-3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| GATA-3 | |
| Monoclonal | |
| Biotin | |
| 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
| Recombinant full-length human GATA-3 protein | |
| 0.1 mL | |
| Cancer, Innate Immunity | |
| 2625 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rGATA3/3870 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction