Learn More
Gata-3 Monoclonal Antibody (TWAJ), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
€ 168.00 - € 409.00
Specifications
Antigen | Gata-3 |
---|---|
Clone | TWAJ |
Concentration | 5 μL/Test |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15580317
|
Affymetrix eBioscience
53-9966-41 |
25 Tests |
€ 168.00
25 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15590317
|
Affymetrix eBioscience
53-9966-42 |
100 Tests |
€ 409.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The monoclonal antibody TWAJ recognizes mouse and human Gata-3, a member of the Gata family of transcription factors. Gata-3 is a T cell-specific transcription factor important for thymic development and Th2 differentiation. Expression during embryonic development is found in the central nervous system, skin, mammary glands and kidney. During development, the expression of Gata-3 is essential as homozygous knock-out of Gata-3 is embryonic lethal. The Gata-3 is also essential for T cell commitment and survival. In the thymus, expression is found mainly on the CD4 single positive cells. During Th2 differentiation, Gata-3 binds to the IL-4 promoter as well as represses the expression of T-bet, thus inhibiting Th1 differentiation. Alternative splice variants have been reported especially in the MCF7 cell line. The TWAJ Human/Mouse Gata-3 antibody will recognize both forms (50 and 45 kDa) of the protein. Staining with the TWAJ Human/Mouse Gata-3 antibody requires the use of the Foxp3/Transcription Factor Staining Buffer Set.(cat 00-5523) Crossreactivity in rhesus monkeys has been published. Applications Reported: This TWAJ antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This TWAJ antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of mouse thymocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol.
Gata-3 Monoclonal antibody specifically detects Gata-3 in Human, Mouse, Porcine, Rhesus Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
Gata-3 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3 | |
GATA3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
GATA3 |
TWAJ | |
Flow Cytometry, Immunocytochemistry | |
Alexa Fluor 488 | |
Rat | |
Human, Mouse, Porcine, Rhesus Monkey | |
P23771, P23772 | |
14462, 2625, 713840, 733631 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.