missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-4 Antibody (Evan), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43112MFV500
This item is not returnable.
View return policy
Description
GATA-4 Monoclonal antibody specifically detects GATA-4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GATA-4 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Evan | |
| Western Blot | |
| GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
| The immunogen for this antibody was Gata4. | |
| 0.1 mL | |
| Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 2626 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction