Learn More
GATA3 Monoclonal Antibody (OTI11H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809113
Encompass_Preferred
 View more versions of this product
Product Code. 17561014
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
GATA3 is abundantly expressed in the T-lymphocyte lineage and is thought to participate in T-cell receptor gene activation through binding to enhancers. The human gene comprises 6 exons distributed over 17 kb of DNA. Its 2 zinc fingers are encoded by 2 separate exons highly conserved with those of GATA1, but no other structural homologies between the 2 genes could be found. The genes for all 4 subunits of the T-cell antigen receptor (alpha, beta, gamma and delta) are controlled by distinct enhancers and their enhancer-binding proteins. Marine and Winoto (1991) identified a common TCR regulatory element by demonstrating binding of the enhancer-binding protein GATA3 to the enhancer elements of all 4 TCR genes. GATA3 had been shown in the chicken to be an enhancer-binding protein containing a zinc finger domain. GATA3 mRNA was demonstrated by Northern blot analysis in T cells but not in B cells or macrophages.
Specifications
Specifications
GATA3
Monoclonal
Unconjugated
GATA3
HDR, HDRS
Mouse
Affinity Chromatography
RUO
2625
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI11H3
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P23771
GATA3
Human recombit protein fragment corresponding to amino acids 155-443 of human GATA3 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b