missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA3 Monoclonal Antibody (OTI11H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809113S
This item is not returnable.
View return policy
Description
GATA3 Monoclonal antibody specifically detects GATA3 in Human samples. It is validated for Western BlotSpecifications
| GATA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11474 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2101 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI11H3 | |
| Unconjugated | |
| ESRRA | |
| EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3 | |
| Human recombinant protein fragment corresponding to amino acids 206-417 of human ESRRA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction