Learn More
GATA3 Monoclonal Antibody (OTI1A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809097S
Encompass_Preferred
 View more versions of this product
Product Code. 17560974
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
GATA3 Monoclonal antibody specifically detects GATA3 in Human samples. It is validated for Western Blot
Specifications
Specifications
GATA3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P23771
Mouse
Affinity chromatography
RUO
2625
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI1A5
Unconjugated
GATA3
EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3
Human recombinant protein fragment corresponding to amino acids 155-443 of human GATA3 produced in E.coli.
30 μL