Learn More
Invitrogen™ Human GATA3 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP1006
Additional Details : Weight : 0.01000kg
Product Code. 15225009
  • € 130.00 / 50µg
Estimated Shipment: 15-04-2024
to see stock.
Description
Description
GATA3 Synthetic Peptide, PEP-1006, from Invitrogen.
This peptide corresponds to 17 amino acids near the carboxy terminus of human GATA3.PEP-1006 can be used as a blocking peptide with polyclonal antibody PA5-20892.
Specifications
Specifications
2625
purified
Human GATA3
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
P23771
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
GATA3
0.02% sodium azide