missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511438
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
Specifications
| GATA4 | |
| Polyclonal | |
| Unconjugated | |
| GATA4 | |
| ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 2626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 2.0 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P43694 | |
| GATA4 | |
| KLH conjugated synthetic peptide between 298-328 amino acids from the C-terminal region of human GATA4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction