Learn More
Invitrogen™ GATA4 Recombinant Rat Monoclonal Antibody (eBioEvan)
Rat Recombinant Monoclonal Antibody
€ 217.00 - € 440.00
Specifications
| Antigen | GATA4 |
|---|---|
| Clone | eBioEvan |
| Concentration | 1.0 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30245302
|
Invitrogen™
740049T |
100 μL |
€ 440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30245423
|
Invitrogen™
740049T20UG |
20 μL |
€ 217.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.Specifications
| GATA4 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P43694, Q08369 | |
| 14463, 2626 | |
| Primary | |
| Affinity chromatography |
| eBioEvan | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human, Mouse | |
| GATA4 | |
| ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
| GATA4 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.