Learn More
GATAD1 Monoclonal Antibody (OTI1G10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804856S
Encompass_Preferred
 View more versions of this product
Product Code. 17564593
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
GATAD1 Monoclonal antibody specifically detects GATAD1 in Human samples. It is validated for Western Blot
Specifications
Specifications
GATAD1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q92570
Mouse
Affinity chromatography
RUO
8013
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1G10
Unconjugated
NR4A3
2310031E19Rik; 2810047M21Rik; 8430439A17Rik; 9130430G15Rik; B330017N08Rik; CMD2B; ENSMUSG00000073241; GATA zinc finger domain containing 1; GATA zinc finger domain-containing 1-like protein; GATA zinc finger domain-containing protein 1; GATA zinc finger domain-containing protein 1-like protein; Gatad1; im:7150682; ocular development associated; ocular development-associated gene protein; ODAG; RG083M05.2; RG83M5.2; si:dkey-231l1.7; Unknown (protein for MGC:136582); zgc:136582
Human recombinant protein fragment corresponding to amino acids 332-626 of human NR4A3 produced in E.coli.