missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GBA2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA572342

Product Code. 15901015

  • € 514.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GBA2 Polyclonal Antibody for Western Blot

Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GBA2
Polyclonal
Unconjugated
GBA2
3.2.1.45; AD035; Beta-glucocerebrosidase 2; beta-glucosidase 2; bile acid; bile acid beta-glucosidase; Bile acid beta-glucosidase GBA2; Bile acid glucosyl transferase GBA2; Cholesterol glucosyltransferase GBA2; Cholesteryl-beta-glucosidase GBA2; F630034E04; GBA2; glucosidase beta 2; glucosidase, beta (bile acid) 2; glucosylceramidase 2; glucosylceramidase beta 2; Kiaa1605; NLGase; non-lysosomal glucosylceramidase; SPG46
Rabbit
Protein A, Antigen affinity chromatography
RUO
230101, 57704
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q69ZF3, Q9HCG7
GBA2
KLH conjugated synthetic peptide between 593-623 amino acids from the Central region of human GBA2.
200 μL
Primary
Human, Mouse
Antibody
IgG
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.