missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBAS Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-77339CL1
This item is not returnable.
View return policy
Description
GBAS Polyclonal antibody specifically detects GBAS in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| GBAS | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| glioblastoma amplified sequence, Glioblastoma-amplified sequence, NIPSNAP2NipSnap2,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2, protein NipSnap homolog 2 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human NIPSNAP2. The immunogen is located within the first 50 amino acids of NIPSNAP2. Amino Acid Squence: RTWTSSSNRSREDS | |
| 0.1 mL | |
| Neuroscience | |
| 2631 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction