missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBE1 Antibody (OTI1D11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70810
This item is not returnable.
View return policy
Description
GBE1 Monoclonal specifically detects GBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.
Specifications
| GBE1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50-100, Immunoprecipitation 2 μg/500 μL, CyTOF-ready | |
| 1,4-alpha-glucan-branching enzyme, amylo-(1,4 to 1,6) transglucosidase, amylo-(1,4 to 1,6) transglycosylase, Brancher enzyme, EC 2.4.1.18, GBE, glucan (1,4-alpha), branching enzyme 1, glycogen branching enzyme, Glycogen-branching enzyme | |
| Mouse | |
| 80.3 kDa | |
| 100 μg | |
| 2632 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF | |
| OTI1D11 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| GBE1 | |
| Full length human recombinant protein of human GBE1 (NP_000149) produced in HEK293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction