missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBE1 Antibody (OTI1D11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70810DL594
This item is not returnable.
View return policy
Description
GBE1 Monoclonal antibody specifically detects GBE1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| GBE1 | |
| Monoclonal | |
| DyLight 594 | |
| 1,4-alpha-glucan-branching enzyme, amylo-(1,4 to 1,6) transglucosidase, amylo-(1,4 to 1,6) transglycosylase, Brancher enzyme, EC 2.4.1.18, GBE, glucan (1,4-alpha), branching enzyme 1, glycogen branching enzyme, Glycogen-branching enzyme | |
| Full length recombinant protein of human GBE1 (NP_000149) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| OTI1D11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 2632 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction