missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBE1 Mouse anti-Human, Clone: OTI5B7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500802
This item is not returnable.
View return policy
Description
GBE1 is a glycogen branching enzyme that catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on the same or a neighboring glycogen chain. Branching of the chains is essential to increase the solubility of the glycogen molecule and, consequently, in reducing the osmotic pressure within cells. Highest level of this enzyme are found in liver and muscle. Mutations in this gene are associated with glycogen storage disease IV (also known as Andersen′s disease).Specifications
| GBE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q04446 | |
| GBE1 | |
| Full length human recombit protein of human GBE1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| OTI5B7 | |
| Unconjugated | |
| GBE1 | |
| APBD, GBE, GSD4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2632 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction