missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GBE1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15796111 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15796111 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796111 Supplier Invitrogen™ Supplier No. PA558236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SVLTPFTPVI DRGIQLHKMI RLITHGLGGE GYLNFMGNEF GHPEWLDFPR KGNNESYHYA RRQFHLTDDD LLRYKFLNNF DRDMNRLEER YGWLAA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

GBE1 is a glycogen branching enzyme that catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on the same or a neighboring glycogen chain. Branching of the chains is essential to increase the solubility of the glycogen molecule and, consequently, in reducing the osmotic pressure within cells. Highest level of this enzyme are found in liver and muscle. Mutations in this gene are associated with glycogen storage disease IV (also known as Andersen's disease).
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen GBE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GBE1
Gene Accession No. Q04446, Q9D6Y9
Gene Alias 1,4-alpha-glucan-branching enzyme; 2310045H19Rik; 2810426P10Rik; amylo-(1,4 to 1,6) transglucosidase; amylo-(1,4 to 1,6) transglycosylase; APBD; Brancher enzyme; D16Ertd536e; GBE; GBE1; glucan (1,4-alpha-), branching enzyme 1; glycogen branching enzyme; glycogen-branching enzyme; GSD4
Gene Symbols GBE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GBE1. Recombinant protein control fragment (Product #RP-95611).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2632, 288333, 74185
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.