missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | GBE1 |
|---|---|
| Concentration | 0.16 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819844
|
Proteintech
20313-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809844
|
Proteintech
20313-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GBE1 is a glycogen branching enzyme that catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on the same or a neighboring glycogen chain. Branching of the chains is essential to increase the solubility of the glycogen molecule and, consequently, in reducing the osmotic pressure within cells. Highest level of this enzyme are found in liver and muscle. Mutations in this gene are associated with glycogen storage disease IV (also known as Andersen′s disease).Specifications
| GBE1 | |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q04446, Q9D6Y9 | |
| 2632, 288333, 74185 | |
| GBE1 Fusion Protein Ag14154 | |
| Primary | |
| -20°C | |
| GBE1 |
| 0.16 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Brancher enzyme, GBE, GBE1, Glycogen branching enzyme | |
| Gbe1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title