missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBF1, Mouse anti-Human, Clone: 4B2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008729-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: THEGTAEGMENMADAVTHARFVGTDPASDEVVLMKILQVLRTLLLTPVGAHLTNESVCEIMQSCFRICFEMRLSELLRKSAEHTLVDMVQLLFTRLPQFKEEPKNYVGTSpecifications
| GBF1 | |
| Monoclonal | |
| Unconjugated | |
| golgi-specific brefeldin A resistant guanine nucleotide exchange factor 1 | |
| ARF1GEF/FLJ21263/FLJ21500/KIAA0248/MGC134877/MGC134878 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4B2 | |
| In 1x PBS, pH 7.4 | |
| NM_004193 | |
| GBF1 | |
| GBF1 (NP_004184.1, 101 a.a. ∼ 209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8729 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction