missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBGT1 Polyclonal Antibody

GBGT1 Polyclonal Antibody
Brand: Thermo Scientific PA5116392
This item is not returnable.
View return policy
Description
GBGT1 Polyclonal Antibody for Western Blot
Specifications
| GBGT1 | |
| Polyclonal | |
| Unconjugated | |
| GBGT1 | |
| A3GALNT; Fgs; Forssman glycolipid synthase; forssman glycolipid synthase-like protein; forssman glycolipid synthetase; Forssman glycolipid synthetase (FS); FS; Gbgt1; Globoside alpha-1,3-N-acetylgalactosaminyltransferase 1; LOC499764; similar to globoside alpha-1,3-N-acetylgalactosaminyltransferase 1; UNQ2513; UNQ2513/PRO6002 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 26301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8N5D6 | |
| GBGT1 | |
| KLH conjugated synthetic peptide between 154-185 amino acids from the Central region of human GBGT1. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction