Learn More
GBL Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-98484CL1
Code produit. 30157857
  • € 822.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
GBL Polyclonal antibody specifically detects GBL in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
GBL
Polyclonal
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
CoraFluor 1
G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1
Produced in rabbits immunized with E. coli-derived Human GBL fragment.
0.1 mL
Cancer, Hypoxia, mTOR Pathway
64223
Store at 4°C in the dark. Do not freeze.