missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00230APC
This item is not returnable.
View return policy
Description
GBP2 Polyclonal antibody specifically detects GBP2 in Human samples. It is validated for ELISA
Specifications
| APC | |
| Antigen and protein A Affinity-purified | |
| Human | |
| Purified |
| Produced in rabbits immunized with purified, recombinant Human GBP2 (Accession#: AAH73163.1; Met1-Cys588) | |
| 0.1 mL | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction