missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP2 Antibody (OTI5C8), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70812PE
This item is not returnable.
View return policy
Description
GBP2 Monoclonal specifically detects GBP2 in Human samples. It is validated for Flow Cytometry.
Specifications
| GBP2 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| GBP2 | |
| Full length recombinant protein of human GBP2 (NP_004111) produced in HEK293T cells. | |
| 0.1 mL | |
| 2634.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI5C8 | |
| Flow Cytometry | |
| GBP-2, GTP-binding protein 2, Guanine nucleotide-binding protein 2, guanylate binding protein 2, interferon-inducible, huGBP-2, interferon-induced guanylate-binding protein 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction